EMAIL THIS PAGE TO A FRIEND
8.41829 EMD Millipore

Diphenyl phosphate

Diphenyl phosphate for synthesis. CAS 838-85-7, molar mass 250.19 g/mol., for synthesis

  • CAS Number 838-85-7

  • Linear Formula C₁₂H₁₁O₄P

  • Molecular Weight 250.19 g/mol

  •  EC Number 212-657-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8418290005)
mp   67 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 66 °C
  Melting range (upper value): ≤ 70 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2919 90 00
RTECS 
TC5470000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?