EMAIL THIS PAGE TO A FRIEND
8.41830 EMD Millipore

1-Decyne

1-Decyne for synthesis. CAS 764-93-2, molar mass 138.25 g/mol., for synthesis

  • CAS Number 764-93-2

  • Linear Formula C₁₀H₁₈

  • Molecular Weight 138.25 g/mol

  •  EC Number 212-132-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8418300025)
bp   174 - 176 °C
density   0.764 - 0.770 g/cm3 (20 °C)
flash point   48 °C
mp   -44 °C
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.764 - 0.770
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2901 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 3295 , 3, II
Declaration (transport by air) IATA-DGR 
UN 3295 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 3295 , 3, II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?