• USA Home
  • 8.41833 - Isobutyl isobutyrate

EMAIL THIS PAGE TO A FRIEND
8.41833 EMD Millipore

Isobutyl isobutyrate

Isobutyl isobutyrate for synthesis. CAS 97-85-8, pH 7 (H₂O)., for synthesis

Synonym: Isobutyric acid isobutyl ester

  • CAS Number 97-85-8

  • Linear Formula C₈H₁₆O₂

  • Molecular Weight 144.21 g/mol

  •  EC Number 202-612-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   147 - 149 °C (1013 hPa)
expl. lim.     1.2 %(V)
vapor pressure   5.76 hPa (25 °C)
solubility   1 g/l
density   0.855 g/cm3 (20 °C)
flash point   38 °C
mp   -81 °C
pH   7 (H₂O)
LD 50 oral   LD50 Rat 12800 mg/kg
LD 50 dermal   LD50 Rabbit > 8600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.853 - 0.857
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 60 19
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
NQ5250000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 2528 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2528 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2528 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?