EMAIL THIS PAGE TO A FRIEND
8.41835 EMD Millipore

1,9-Nonanediol

1,9-Nonanediol for synthesis. CAS 3937-56-2, pH (9 g/l, H₂O, 20 °C) neutral., for synthesis

Synonym: Nonamethylene glycol

  • CAS Number 3937-56-2

  • Linear Formula C₉H₂₀O₂

  • Molecular Weight 160.25 g/mol

  •  EC Number 223-517-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8418350025)
vapor pressure   <0.01 hPa (20 °C)
solubility   9 g/l
density   0.918 g/cm3 (50 °C)
flash point   162 °C
autoignition temp.   280 °C
mp   45 - 46 °C
LD 50 oral   LD50 Rat > 3000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 45 °C
  Melting range (upper value): ≤ 48 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 39 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?