• USA Home
  • 8.41836 - 2-Fluoro-5-nitroaniline

EMAIL THIS PAGE TO A FRIEND
8.41836 EMD Millipore

2-Fluoro-5-nitroaniline

2-Fluoro-5-nitroaniline for synthesis. CAS 369-36-8, molar mass 156.12 g/mol., for synthesis

  • CAS Number 369-36-8

  • Linear Formula C₆H₅FN₂O₂

  • Molecular Weight 156.12 g/mol

  •  EC Number 206-720-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8418360005)
mp   101 - 104 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 100 °C
  Melting range (upper value): ≤ 104 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Storage class 
4.1B Flammable solid hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 1325 , 4.1, II
Declaration (transport by air) IATA-DGR 
UN 1325 , 4.1, II
Declaration (transport by sea) IMDG-Code 
UN 1325 , 4.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?