• USA Home
  • 8.41838 - DL-Methyl mandelate

EMAIL THIS PAGE TO A FRIEND
8.41838 EMD Millipore

DL-Methyl mandelate

DL-Methyl mandelate for synthesis. CAS 4358-87-6, molar mass 166.18 g/mol., for synthesis

  • CAS Number 4358-87-6

  • Linear Formula C₉H₁₀O₃

  • Molecular Weight 166.18 g/mol

  •  EC Number 224-434-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8418380100)
flash point   113 °C
mp   54 - 56 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 54 °C
  Melting range (upper value): ≤ 57 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 19 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?