• USA Home
  • 8.41850 - 1,12-Dibromododecane

EMAIL THIS PAGE TO A FRIEND
8.41850 EMD Millipore

1,12-Dibromododecane

1,12-Dibromododecane for synthesis. CAS 3344-70-5, molar mass 328.12 g/mol., for synthesis

  • CAS Number 3344-70-5

  • Linear Formula C₁₂H₂₄Br₂

  • Molecular Weight 328.12 g/mol

  •  EC Number 222-096-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8418500005)
flash point   >110 °C
mp   37 - 39 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Melting range (lower value): ≥ 37 °C
  Melting range (upper value): ≤ 40 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
RTECS 
JR2295000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?