• USA Home
  • 8.41852 - 2,3,5-Triiodobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.41852 EMD Millipore

2,3,5-Triiodobenzoic acid

2,3,5-Triiodobenzoic acid for synthesis. CAS 88-82-4, molar mass 499.81 g/mol., for synthesis

Synonym: TIBA

  • CAS Number 88-82-4

  • Linear Formula C₇H₃I₃O₂

  • Molecular Weight 499.81 g/mol

  •  EC Number 201-859-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8418520005)
mp   226 °C
LD 50 oral   LD50 Rat 813 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H302: Harmful if swallowed.
RTECS 
DH9275000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?