• USA Home
  • 8.41854 - Tetramethylthiourea

EMAIL THIS PAGE TO A FRIEND
8.41854 EMD Millipore

Tetramethylthiourea

Tetramethylthiourea for synthesis. CAS 2782-91-4, molar mass 132.22 g/mol., for synthesis

  • CAS Number 2782-91-4

  • Linear Formula C₅H₁₂N₂S

  • Molecular Weight 132.22 g/mol

  •  EC Number 220-488-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8418540025)
bp   245 °C (1013 hPa)
vapor pressure   1.9 hPa (25 °C)
solubility   32 g/l
flash point   100 °C
mp   75 - 78 °C
LD 50 oral   LD50 Rat 920 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 75 °C
  Melting range (upper value): ≤ 78 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H302: Harmful if swallowed.
RTECS 
YU2750000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?