• USA Home
  • 8.41860 - 1-Aminocyclohexanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41860 EMD Millipore

1-Aminocyclohexanecarboxylic acid

1-Aminocyclohexanecarboxylic acid for synthesis. CAS 2756-85-6, molar mass 143.18 g/mol., for synthesis

  • CAS Number 2756-85-6

  • Linear Formula C₇H₁₃NO₂

  • Molecular Weight 143.18 g/mol

  •  EC Number 220-411-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8418600010)
mp   >300 °C
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
RTECS 
GU8393000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?