EMAIL THIS PAGE TO A FRIEND
8.41868 EMD Millipore

2-Bromoacetamide

2-Bromoacetamide for synthesis. CAS 683-57-8, chemical formula BrCH₂CONH₂., for synthesis

  • CAS Number 683-57-8

  • Linear Formula C₂H₄BrNO

  • Molecular Weight 137.96 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
solubility   160 g/l
mp   87 - 90 °C
LD 50 oral   LD50 Rat 100 mg/kg
LD 50 dermal   LD50 Rat 3160 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 87 °C
  Melting range (upper value): ≤ 90 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
AB4587000
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2923 , 8 (6.1), II
Declaration (transport by air) IATA-DGR 
UN 2923 , 8 (6.1), II
Declaration (transport by sea) IMDG-Code 
UN 2923 , 8 (6.1), II
Categories of danger 
toxic, corrosive
R Phrase 
R 25-34
Toxic if swallowed.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?