• USA Home
  • 8.41872 - 2-Methyl-1,3-dioxolane

EMAIL THIS PAGE TO A FRIEND
8.41872 EMD Millipore

2-Methyl-1,3-dioxolane

2-Methyl-1,3-dioxolane for synthesis. CAS 497-26-7, molar mass 88.11 g/mol., for synthesis

Synonym: Acetaldehyde ethylene acetal

  • CAS Number 497-26-7

  • Linear Formula C₄H₈O₂

  • Molecular Weight 88.11 g/mol

  •  EC Number 207-841-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8418720010)
expl. lim.     1.8 %(V)
vapor pressure   57.3 hPa (20 °C)
density   0.982 g/cm3 (20 °C)
flash point   -2 °C
LD 50 oral   LD50 Rat 2900 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.980 - 0.984
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P403 + P235: Store in a well-ventilated place. Keep cool.
RTECS 
JI3509000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 25
Avoid contact with eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?