• USA Home
  • 8.41883 - 2-Phenylquinoline-4-caboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41883 EMD Millipore

2-Phenylquinoline-4-caboxylic acid

2-Phenylquinoline-4-caboxylic acid for synthesis. CAS 132-60-5, molar mass 249.26 g/mol., for synthesis

  • CAS Number 132-60-5

  • Linear Formula C₁₆H₁₁NO₂

  • Molecular Weight 249.26 g/mol

  •  EC Number 205-067-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   213 - 216 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 213 °C
  Melting range (upper value): ≤ 216 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 49 10
RTECS 
GD4025000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?