• USA Home
  • 8.41886 - 1-(3,4-Dichlorophenyl)-piperazine

EMAIL THIS PAGE TO A FRIEND
8.41886 EMD Millipore

1-(3,4-Dichlorophenyl)-piperazine

1-(3,4-Dichlorophenyl)-piperazine for synthesis. CAS 57260-67-0, molar mass 231.13 g/mol., for synthesis

  • CAS Number 57260-67-0

  • Linear Formula C₁₀H₁₂Cl₂N₂

  • Molecular Weight 231.13 g/mol

  •  EC Number 260-652-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8418860010)
bp   179 °C (20 hPa)
mp   59.0 - 65.0 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.5 % (a/a)
  Melting range (lower value): ≥ 59 °C
  Melting range (upper value): ≤ 65 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?