• USA Home
  • 8.42126 - Cyclohexane-1,4-dicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.42126 EMD Millipore

Cyclohexane-1,4-dicarboxylic acid

Cyclohexane-1,4-dicarboxylic acid (mixture of isomeres) for synthesis. CAS 1076-97-7, molar mass 172.18 g/mol., (mixture of isomeres) for synthesis

  • CAS Number 1076-97-7

  • Linear Formula C₈H₁₂O₄

  • Molecular Weight 172.18 g/mol

  •  EC Number 214-068-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8421260100)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 165 °C
  Melting range (upper value): ≤ 168 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 20 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
GU9060000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?