• USA Home
  • 8.42251 - 2-(4-Chlorobenzoyl) benzoic acid

EMAIL THIS PAGE TO A FRIEND
8.42251 EMD Millipore

2-(4-Chlorobenzoyl) benzoic acid

2-(4-Chlorobenzoyl) benzoic acid for synthesis. CAS 85-56-3, pH 3 (1 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 85-56-3

  • Linear Formula C₁₄H₉ClO₃

  • Molecular Weight 260.7 g/mol

  •  EC Number 201-615-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
solubility   0.28 g/l
flash point   >300 °C
autoignition temp.   >600 °C DIN 51794
mp   148 - 151 °C
pH   3 (1 g/l, H₂O, 20 °C)
bulk density   500 kg/m3
LD 50 oral   LD50 Rat 1500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 148 °C
  Melting range (upper value): ≤ 151 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?