• USA Home
  • 8.42274 - 2,2,3,3-Tetrafluoro-1-propanol

EMAIL THIS PAGE TO A FRIEND
8.42274 EMD Millipore

2,2,3,3-Tetrafluoro-1-propanol

2,2,3,3-Tetrafluoro-1-propanol for synthesis. CAS 76-37-9, molar mass 132.06 g/mol., for synthesis

  • CAS Number 76-37-9

  • Linear Formula C₃H₄F₄O

  • Molecular Weight 132.06 g/mol

  •  EC Number 200-955-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8422740100)
bp   107 - 109 °C (1013 hPa)
density   1.485 - 1.490 g/cm3 (20 °C)
flash point   46 °C
mp   -15 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.485 - 1.490
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 59 98
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H332: Harmful if inhaled.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
UB9400000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable, harmful
R Phrase 
R 10-20
Flammable.Harmful by inhalation.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?