EMAIL THIS PAGE TO A FRIEND
8.42309 EMD Millipore

2,3-Dihydrofuran

2,3-Dihydrofuran for synthesis. CAS 1191-99-7, molar mass 70.09 g/mol., for synthesis

  • CAS Number 1191-99-7

  • Linear Formula C₄H₆O

  • Molecular Weight 70.09 g/mol

  •  EC Number 214-747-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8423090100)
bp   54 - 55 °C (1013 hPa)
vapor pressure   190 hPa (20 °C)
density   0.93 g/cm3 (20 °C)
flash point   -24 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Water (GC): ≤ 0.30 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 19 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H319: Causes serious eye irritation.
EUH019: May form explosive peroxides.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3271 , 3, II
Declaration (transport by air) IATA-DGR 
UN 3271 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 3271 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-19-36
Highly flammable.May form explosive peroxides.Irritating to eyes.
S Phrase 
S 16-26
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?