• USA Home
  • 8.42351 - 2,4-Dichlorobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.42351 EMD Millipore

2,4-Dichlorobenzotrifluoride

2,4-Dichlorobenzotrifluoride for synthesis. CAS 320-60-5, molar mass 215 g/mol., for synthesis

  • CAS Number 320-60-5

  • Linear Formula C₇H₃Cl₂F₃

  • Molecular Weight 215 g/mol

  •  EC Number 206-278-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8423510010)
bp   117 - 118 °C (1013 hPa)
vapor pressure   1.48 hPa (25 °C)
density   1.490 g/cm3 (20 °C)
flash point   72 °C
LD 50 oral   LD50 Rat 2150 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.488 - 1.492
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
CZ5566877
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?