• USA Home
  • 8.42413 - 2,6-Dichloro-4-methylnicotinonitrile

EMAIL THIS PAGE TO A FRIEND
8.42413 EMD Millipore

2,6-Dichloro-4-methylnicotinonitrile

2,6-Dichloro-4-methylnicotinonitrile for synthesis. CAS 875-35-4, pH 3.5 (H₂O, 20 °C) (aqueous suspension)., for synthesis

  • CAS Number 875-35-4

  • Linear Formula C₇H₄Cl₂N₂

  • Molecular Weight 187.03 g/mol

  •  EC Number 212-873-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
solubility   0.4 g/l
autoignition temp.   >600 °C DIN 51794
pH   3.5 (H₂O, 20 °C) (aqueous suspension)
bulk density   500 kg/m3
LD 50 oral   LD50 Rat 3600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 110 °C
  Melting range (upper value): ≤ 112 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Categories of danger 
dangerous for the environment
R Phrase 
R 52/53
Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?