• USA Home
  • 8.42447 - 1-Bromo-2-methoxyethane

EMAIL THIS PAGE TO A FRIEND
8.42447 EMD Millipore

1-Bromo-2-methoxyethane

1-Bromo-2-methoxyethane (stabilised) for synthesis. CAS 6482-24-2, molar mass 138.99 g/mol., (stabilised) for synthesis

  • CAS Number 6482-24-2

  • Linear Formula C₃H₇BrO

  • Molecular Weight 138.99 g/mol

  •  EC Number 229-339-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8424470025)
bp   107 - 110 °C (1013 hPa)
density   1.46 g/cm3
flash point   28 °C
autoignition temp.   200 °C DIN 51794
LD 50 oral   LD50 Rat > 200 - 400 mg/kg
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.456 - 1.466
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 19 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H301: Toxic if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1992 , 3 (6.1), III
Declaration (transport by air) IATA-DGR 
UN 1992 , 3 (6.1), III
Declaration (transport by sea) IMDG-Code 
UN 1992 , 3 (6.1), III
Categories of danger 
flammable, harmful, irritant
R Phrase 
R 10-22-36/37/38
Flammable.Harmful if swallowed.Irritating to eyes, respiratory system and skin.
S Phrase 
S 16-26
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?