EMAIL THIS PAGE TO A FRIEND
8.42513 EMD Millipore

2-Etylhexanal

2-Etylhexanal for synthesis. CAS 123-05-7, molar mass 128.22 g/mol., for synthesis

  • CAS Number 123-05-7

  • Linear Formula C₈H₁₆O

  • Molecular Weight 128.22 g/mol

  •  EC Number 204-596-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8425130050)
bp   162 °C (1013 hPa)
expl. lim.     1.0 - 6.6 %(V)
vapor pressure   2 hPa (20 °C)
solubility   0.01 g/l
density   0.82 g/cm3 (20 °C)
flash point   45.5 °C DIN 51755 Part 1
autoignition temp.   180 °C DIN 51794
mp   -85 °C
LD 50 oral   LD50 Rat 2732 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.818 - 0.821
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
MN7525000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1191 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1191 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1191 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-38
Flammable.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?