• USA Home
  • 8.42570 - 2-Methoxyethyl cyanoacetate

EMAIL THIS PAGE TO A FRIEND
8.42570 EMD Millipore

2-Methoxyethyl cyanoacetate

2-Methoxyethyl cyanoacetate for synthesis. CAS 10258-54-5, molar mass 143.14 g/mol., for synthesis

  • CAS Number 10258-54-5

  • Linear Formula C₆H₉NO₃

  • Molecular Weight 143.14 g/mol

  •  EC Number 233-597-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8425700100)
  25 ml in Glass bottle (8425700025)
bp   110 °C (1.1 hPa)
vapor pressure   0.27 hPa (25 °C)
solubility   745 g/l
density   1.125 g/cm3 (25 °C)
flash point   113 °C
autoignition temp.   374 °C
mp   -12.5 °C
LD 50 oral   LD50 Rat 4300 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.125 - 1.130
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
RTECS 
AG4350000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?