EMAIL THIS PAGE TO A FRIEND
8.42600 EMD Millipore

2-Nitrothiophene

2-Nitrothiophene for synthesis. CAS 609-40-5, molar mass 129.14 g/mol., for synthesis

  • CAS Number 609-40-5

  • Linear Formula C₄H₃NO₂S

  • Molecular Weight 129.14 g/mol

  •  EC Number 210-190-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8426000010)
bp   224 - 225 °C (1013 hPa)
flash point   93 °C
mp   43 - 45 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 80.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
RTECS 
XN0030000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?