• USA Home
  • 8.42675 - 3,4-Dimethylanisole

EMAIL THIS PAGE TO A FRIEND
8.42675 EMD Millipore

3,4-Dimethylanisole

3,4-Dimethylanisole for synthesis. CAS 4685-47-6, molar mass 136.19 g/mol., for synthesis

  • CAS Number 4685-47-6

  • Linear Formula C₉H₁₂O

  • Molecular Weight 136.19 g/mol

  •  EC Number 225-142-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8426750050)
flash point   76 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.968 - 0.972
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?