EMAIL THIS PAGE TO A FRIEND
8.42746 EMD Millipore

3-Bromo-o-xylene

3-Bromo-o-xylene for synthesis. CAS 576-23-8, molar mass 185.06 g/mol., for synthesis

  • CAS Number 576-23-8

  • Linear Formula C₈H₉Br

  • Molecular Weight 185.06 g/mol

  •  EC Number 209-398-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8427460005)
bp   212 °C (1013 hPa)
density   1.365 g/cm3 (20 °C)
flash point   80 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.370 - 1.375
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1701 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 1701 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 1701 , 6.1, II, Segregation Group: 10 (Liquid halogenated hydrocarbons)
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?