• USA Home
  • 8.42770 - 3-Ethoxypropylamine

EMAIL THIS PAGE TO A FRIEND
8.42770 EMD Millipore

3-Ethoxypropylamine

3-Ethoxypropylamine for synthesis. CAS 6291-85-6, pH 12 (100 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 6291-85-6

  • Linear Formula C₅H₁₃NO

  • Molecular Weight 103.17 g/mol

  •  EC Number 228-545-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8427700100)
expl. lim.     0.8 - 8.1 %(V)
vapor pressure   6.9 hPa (20 °C)
density   0.86 g/cm3 (20 °C)
flash point   33 °C DIN 51755 Part 1
autoignition temp.   245 °C DIN 51794
mp   <-70 °C
pH   12 (100 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 1115 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.853 - 0.857
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302 + H332: Harmful if swallowed or if inhaled
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
UI2700000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2734 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2734 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2734 , 8 (3), II, Segregation Group: 18 (Alkalis)
Categories of danger 
flammable, harmful, corrosive
R Phrase 
R 10-20/22-34
Flammable.Harmful by inhalation and if swallowed.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?