• USA Home
  • 8.42792 - 3'-Hydroxyacetophenone

EMAIL THIS PAGE TO A FRIEND
8.42792 EMD Millipore

3'-Hydroxyacetophenone

3'-Hydroxyacetophenone for synthesis. CAS 121-71-1, molar mass 136.15 g/mol., for synthesis

  • CAS Number 121-71-1

  • Linear Formula C₈H₈O₂

  • Molecular Weight 136.15 g/mol

  •  EC Number 204-494-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8427920025)
bp   296 °C (1013 hPa)
vapor pressure   1 Pa (25 °C)
solubility   22 g/l
density   1.1 g/cm3
mp   94 - 97 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 94 °C
  Melting range (upper value): ≤ 97 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 50 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?