• USA Home
  • 8.42985 - 4-Diethylaminobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.42985 EMD Millipore

4-Diethylaminobenzaldehyde

4-Diethylaminobenzaldehyde for synthesis. CAS 120-21-8, molar mass 177.25 g/mol., for synthesis

  • CAS Number 120-21-8

  • Linear Formula C₁₁H₁₅NO

  • Molecular Weight 177.25 g/mol

  •  EC Number 204-377-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8429850010)
bp   174 °C (9 hPa)
solubility   2.5 g/l
density   1.03 g/cm3
flash point   165 °C
autoignition temp.   410 °C
mp   37 - 41 °C
LD 50 oral   LD50 Rat 6400 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 37 °C
  Melting range (upper value): ≤ 41 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 39 00
RTECS 
CU5612850
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?