• USA Home
  • 8.42990 - 4-Ethoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.42990 EMD Millipore

4-Ethoxybenzoic acid

4-Ethoxybenzoic acid for synthesis. CAS 619-86-3, molar mass 166.18 g/mol., for synthesis

  • CAS Number 619-86-3

  • Linear Formula C₉H₁₀O₃

  • Molecular Weight 166.18 g/mol

  •  EC Number 210-616-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8429900100)
vapor pressure   0.07 Pa (25 °C)
mp   197 - 199 °C
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 197 °C
  Melting range (upper value): ≤ 200 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?