• USA Home
  • 8.43049 - 4-Methyl-3-nitroaniline

EMAIL THIS PAGE TO A FRIEND
8.43049 EMD Millipore

4-Methyl-3-nitroaniline

4-Methyl-3-nitroaniline for synthesis. CAS 119-32-4, molar mass 152.15 g/mol., for synthesis

  • CAS Number 119-32-4

  • Linear Formula C₇H₈N₂O₂

  • Molecular Weight 152.15 g/mol

  •  EC Number 204-314-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8430490025)
solubility   <1 g/l
flash point   157 °C
mp   76 - 78 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 76 °C
  Melting range (upper value): ≤ 80 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 43 00
Hazard Statement(s) 
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
H373: May cause damage to organs through prolonged or repeated exposure.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
XU8227000
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
toxic, dangerous for the environment
R Phrase 
R 23/24/25-33-51/53
Toxic by inhalation, in contact with skin and if swallowed.Danger of cumulative effects.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 28-36/37-45-61
After contact with skin, wash immediately with plenty of soap and water.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?