• USA Home
  • 8.43050 - 4-Methyl-3-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.43050 EMD Millipore

4-Methyl-3-nitrobenzoic acid

4-Methyl-3-nitrobenzoic acid for synthesis. CAS 96-98-0, molar mass 181.15 g/mol., for synthesis

  • CAS Number 96-98-0

  • Linear Formula C₈H₇NO₄

  • Molecular Weight 181.15 g/mol

  •  EC Number 202-549-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8430500100)
  25 g in Glass bottle (8430500025)
vapor pressure   <0.001 Pa (20 °C)
solubility   0.257 g/l
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 188 °C
  Melting range (upper value): ≤ 192 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 22-24/25
Do not breathe dust.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?