EMAIL THIS PAGE TO A FRIEND
8.43176 EMD Millipore

6-Nitroindazole

6-Nitroindazole for synthesis. CAS 7597-18-4, molar mass 163.14 g/mol., for synthesis

  • CAS Number 7597-18-4

  • Linear Formula C₇H₅N₃O₂

  • Molecular Weight 163.14 g/mol

  •  EC Number 231-500-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8431760025)
mp   177 - 180 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 178 °C
  Melting range (upper value): ≤ 182 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Hazard Statement(s) 
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe dust.
P262: Do not get in eyes, on skin, or on clothing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
NK7962100
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36
Irritating to eyes.
S Phrase 
S 22-24
Do not breathe dust.Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?