• USA Home
  • 8.43230 - Sodium benzenesulphinate

EMAIL THIS PAGE TO A FRIEND
8.43230 EMD Millipore

Sodium benzenesulphinate

Sodium benzenesulphinate for synthesis. CAS 873-55-2, molar mass 164.16 g/mol., for synthesis

  • CAS Number 873-55-2

  • Linear Formula C₆H₅NaO₂S

  • Molecular Weight 164.16 g/mol

  •  EC Number 212-842-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8432300100)
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Water (K. F.): ≤ 1.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?