• USA Home
  • 8.43250 - Benzyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
8.43250 EMD Millipore

Benzyl isothiocyanate

Benzyl isothiocyanate for synthesis. CAS 622-78-6, molar mass 149.22 g/mol., for synthesis

  • CAS Number 622-78-6

  • Linear Formula C₈H₇NS

  • Molecular Weight 149.22 g/mol

  •  EC Number 210-753-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8432500005)
bp   243 °C (1013 hPa)
vapor pressure   0.02 hPa (25 °C)
solubility   0.1 g/l
density   1.084 g/cm3 (20 °C)
flash point   106 °C
mp   41 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P342 + P311: If experiencing respiratory symptoms: Call a POISON CENTER/doctor.
RTECS 
NX8250000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2206 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2206 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2206 , 6.1, II
Categories of danger 
toxic, irritant, sensitizing, dangerous for the environment
R Phrase 
R 22-36/37/38-42/43
Harmful if swallowed.Irritating to eyes, respiratory system and skin.May cause sensitisation by inhalation and skin contact.
S Phrase 
S 23-24-37-45
Do not breathe vapour.Avoid contact with skin.Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?