• USA Home
  • 8.43254 - Benzyltriethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
8.43254 EMD Millipore

Benzyltriethylammonium bromide

Benzyltriethylammonium bromide for synthesis. CAS 5197-95-5, molar mass 272.23 g/mol., for synthesis

  • CAS Number 5197-95-5

  • Linear Formula C₁₃H₂₂N*Br

  • Molecular Weight 272.23 g/mol

  •  EC Number 225-986-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8432540025)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric; calculated on dried substance): ≥ 99.0 %
  Loss on drying (Loss on drying at 105°C): ≤ 3.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?