• USA Home
  • 8.43264 - Bis(2-nitrophenyl) disulfide

EMAIL THIS PAGE TO A FRIEND
8.43264 EMD Millipore

Bis(2-nitrophenyl) disulfide

Bis(2-nitrophenyl) disulfide for synthesis. CAS 1155-00-6, molar mass 308.34 g/mol., for synthesis

  • CAS Number 1155-00-6

  • Linear Formula C₁₂H₈N₂O₄S₂

  • Molecular Weight 308.34 g/mol

  •  EC Number 214-581-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8432640100)
  25 g in Glass bottle (8432640025)
density   1.62 g/cm3 (-123.16 °C)
mp   192 - 193 °C
shipped in   ambient
storage temp.   Store at +15°C to +25°C.
specifications   Assay (HPLC, area%): ≥ 95.0 % (a/a)
  Melting range (lower value): ≥ 190 °C
  Melting range (upper value): ≤ 196 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
4.1A Other explosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?