EMAIL THIS PAGE TO A FRIEND
8.43275 EMD Millipore

Calcium propionate

Calcium propionate for synthesis. CAS 4075-81-4, pH 9.2 (200 g/l, H₂O, 20 °C) (IUCLID)., for synthesis

  • CAS Number 4075-81-4

  • Linear Formula C₆H₁₀CaO₄

  • Molecular Weight 186.22 g/mol

  •  EC Number 223-795-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8432750025)
solubility   280 g/l
autoignition temp.   490 °C
mp   382 - 384 °C
pH   9.2 (200 g/l, H₂O, 20 °C) (IUCLID)
bulk density   400 kg/m3
LD 50 oral   LD50 Rat 3920 - 4380 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (complexometric, calc. on anhydrous substance): ≥ 99.0 %
  Water (K. F.): ≤ 9.5 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 50 00
RTECS 
UE8450000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?