• USA Home
  • 8.43342 - Diethylcarbamyl chloride

EMAIL THIS PAGE TO A FRIEND
8.43342 EMD Millipore

Diethylcarbamyl chloride

Diethylcarbamyl chloride for synthesis. CAS 88-10-8, molar mass 135.59 g/mol., for synthesis

  • CAS Number 88-10-8

  • Linear Formula C₅H₁₀ClNO

  • Molecular Weight 135.59 g/mol

  •  EC Number 201-798-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8433420010)
bp   117 - 123 °C (133 hPa)
vapor pressure   1 hPa (20 °C)
density   1.07 g/cm3 (20 °C)
flash point   76 °C
autoignition temp.   400 °C
mp   -32 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Density (d 20 °C/ 4 °C): 1.070 - 1.074
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Hazard Statement(s) 
H302 + H332: Harmful if swallowed or if inhaled
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H351: Suspected of causing cancer.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P313: IF exposed or concerned: Get medical advice/ attention.
RTECS 
FD4025000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, III
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, III, Segregation Group: 1 (Acids)
Categories of danger 
harmful, irritant, carcinogenic
R Phrase 
R 20/22-36/37/38-40
Harmful by inhalation and if swallowed.Irritating to eyes, respiratory system and skin.Limited evidence of a carcinogenic effect.
S Phrase 
S 26-36/37
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?