• USA Home
  • 8.43345 - Dimethoxydimethylsilane

EMAIL THIS PAGE TO A FRIEND
8.43345 EMD Millipore

Dimethoxydimethylsilane

Dimethoxydimethylsilane for synthesis. CAS 1112-39-6, molar mass 120.23 g/mol., for synthesis

  • CAS Number 1112-39-6

  • Linear Formula C₄H₁₂O₂Si

  • Molecular Weight 120.23 g/mol

  •  EC Number 214-189-4

Purchase

Properties

packaging   25 ml in Glass bottle (8433450025)
bp   81 °C (1013 hPa)
density   0.864 g/cm3
flash point   10 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.862 - 0.866
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P403 + P235: Store in a well-ventilated place. Keep cool.
RTECS 
VV3641666
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16-25
Keep away from sources of ignition - No smoking.Avoid contact with eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?