• USA Home
  • 8.43400 - Ethyl 4-nitrobenzoate

EMAIL THIS PAGE TO A FRIEND
8.43400 EMD Millipore

Ethyl 4-nitrobenzoate

Ethyl 4-nitrobenzoate for synthesis. CAS 99-77-4, molar mass 195.17 g/mol., for synthesis

  • CAS Number 99-77-4

  • Linear Formula C₉H₉NO₄

  • Molecular Weight 195.17 g/mol

  •  EC Number 202-786-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8434000025)
bp   320 °C (1013 hPa)
mp   54 - 57 °C
bulk density   380 kg/m3
LD 50 oral   LD50 Rat 2660 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 54 °C
  Melting range (upper value): ≤ 57 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
RTECS 
DH5600000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?