• USA Home
  • 8.43427 - Ethyl trichloroacetate

EMAIL THIS PAGE TO A FRIEND
8.43427 EMD Millipore

Ethyl trichloroacetate

Ethyl trichloroacetate for synthesis. CAS 515-84-4, molar mass 191.44 g/mol., for synthesis

  • CAS Number 515-84-4

  • Linear Formula C₄H₅Cl₃O₂

  • Molecular Weight 191.44 g/mol

  •  EC Number 208-212-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8434270025)
bp   168 °C (1013 hPa)
vapor pressure   1 hPa (20 °C)
solubility   <0.1 g/l
density   1.38 g/cm3 (20 °C)
flash point   85 °C
autoignition temp.   450 °C
LD 50 oral   LD50 Rat 900 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.381 - 1.384
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 40 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-38
Harmful if swallowed.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?