EMAIL THIS PAGE TO A FRIEND
8.43536 EMD Millipore

Methyl carbazate

Methyl carbazate for synthesis. CAS 6294-89-9, molar mass 90.08 g/mol., for synthesis

  • CAS Number 6294-89-9

  • Linear Formula C₂H₆N₂O₂

  • Molecular Weight 90.08 g/mol

  •  EC Number 228-560-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8435360010)
bp   108 °C (12 hPa)
vapor pressure   3 hPa (25 °C)
solubility   578 g/l
mp   70 - 74 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 70 °C
  Melting range (upper value): ≤ 74 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?