• USA Home
  • 8.43570 - N,N'-Bis(3-aminopropyl)ethylendiamine

EMAIL THIS PAGE TO A FRIEND
8.43570 EMD Millipore

N,N'-Bis(3-aminopropyl)ethylendiamine

N,N'-Bis(3-aminopropyl)ethylendiamine for synthesis. CAS 10563-26-5, pH >12 (10 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 10563-26-5

  • Linear Formula C₈H₂₂N₄

  • Molecular Weight 174.29 g/mol

  •  EC Number 234-147-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8435700010)
bp   169 - 175 °C (15 hPa)
vapor pressure   <1 hPa (20 °C)
density   0.95 g/cm3 (20 °C)
flash point   164 °C
autoignition temp.   280 °C
mp   -1.5 °C
pH   >12 (10 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 1140 mg/kg
LD 50 dermal   LD50 Rabbit 190 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.951 - 0.953
  Water (K. F.): ≤ 0.50 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H310: Fatal in contact with skin.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
TX8032950
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2922 , 8 (6.1), II
Declaration (transport by air) IATA-DGR 
UN 2922 , 8 (6.1), II
Declaration (transport by sea) IMDG-Code 
UN 2922 , 8 (6.1), II, Segregation Group: 18 (Alkalis)
Categories of danger 
toxic, corrosive
R Phrase 
R 22-24-34-43
Harmful if swallowed.Toxic in contact with skin.Causes burns.May cause sensitisation by skin contact.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?