• USA Home
  • 8.43583 - N-(2-Aminoethyl)-1,3-propanediamine

EMAIL THIS PAGE TO A FRIEND
8.43583 EMD Millipore

N-(2-Aminoethyl)-1,3-propanediamine

N-(2-Aminoethyl)-1,3-propanediamine for synthesis. CAS 13531-52-7, pH >7 (H₂O, 20 °C)., for synthesis

  • CAS Number 13531-52-7

  • Linear Formula C₅H₁₅N₃

  • Molecular Weight 117.2 g/mol

  •  EC Number 236-882-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8435830025)
expl. lim.     1.0 - 8.1 %(V)
vapor pressure   <1 hPa (20 °C)
density   0.93 g/cm3 (20 °C)
flash point   97 °C DIN 51758
autoignition temp.   325 °C DIN 51794
mp   -11 °C
pH   >7 (H₂O, 20 °C)
LD 50 oral   LD50 Rat 650 mg/kg
LD 50 dermal   LD50 Rabbit 190 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.935 - 0.940
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H310: Fatal in contact with skin.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2922 , 8 (6.1), II
Declaration (transport by air) IATA-DGR 
UN 2922 , 8 (6.1), II
Declaration (transport by sea) IMDG-Code 
UN 2922 , 8 (6.1), II
Categories of danger 
toxic, corrosive
R Phrase 
R 22-24-34
Harmful if swallowed.Toxic in contact with skin.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?