• USA Home
  • 8.43609 - 3-Methylamino propylamine

EMAIL THIS PAGE TO A FRIEND
8.43609 EMD Millipore

3-Methylamino propylamine

3-Methylamino propylamine for synthesis. CAS 6291-84-5, pH 13.5 (100 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 6291-84-5

  • Linear Formula C₄H₁₂N₂

  • Molecular Weight 88.15 g/mol

  •  EC Number 228-544-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8436090025)
expl. lim.     1.8 - 12.8 %(V)
density   0.85 g/cm3 (20 °C)
flash point   42 °C DIN 51755 Part 1
autoignition temp.   305 °C DIN 51794
mp   -72 °C
pH   13.5 (100 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 1063 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.848 - 0.852
  Water (K. F.): ≤ 0.50 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
TX8242500
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2734 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2734 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2734 , 8 (3), II, Segregation Group: 18 (Alkalis)
Categories of danger 
flammable, harmful, corrosive
R Phrase 
R 10-20/21/22-35
Flammable.Harmful by inhalation, in contact with skin and if swallowed.Causes severe burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?