EMAIL THIS PAGE TO A FRIEND
8.43648 EMD Millipore

Phenetole

Phenetole for synthesis. CAS 103-73-1, molar mass 122.17 g/mol., for synthesis

  • CAS Number 103-73-1

  • Linear Formula C₈H₁₀O

  • Molecular Weight 122.17 g/mol

  •  EC Number 203-139-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8436480100)
bp   170 °C (1013 hPa)
vapor pressure   2.27 hPa (25 °C)
solubility   0.569 g/l
density   0.966 g/cm3 (20 °C)
flash point   57 °C
mp   -30 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.963 - 0.967
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
SI7700000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 3271 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3271 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3271 , 3, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?