• USA Home
  • 8.43662 - Phthalimidoacetaldehyd diethylacetal

EMAIL THIS PAGE TO A FRIEND
8.43662 EMD Millipore

Phthalimidoacetaldehyd diethylacetal

Phthalimidoacetaldehyd diethylacetal for synthesis. CAS 78902-09-7, molar mass 263.29 g/mol., for synthesis

  • CAS Number 78902-09-7

  • Linear Formula C₁₄H₁₇NO₄

  • Molecular Weight 263.29 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8436620025)
mp   72 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 69 °C
  Melting range (upper value): ≤ 72 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2925 19 95
Precautionary Statement(s) 
P260: Do not breathe dust.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 22-24/25
Do not breathe dust.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?