• USA Home
  • 8.43677 - Sodiumtoluene-4-sulfinate

EMAIL THIS PAGE TO A FRIEND
8.43677 EMD Millipore

Sodiumtoluene-4-sulfinate

Sodiumtoluene-4-sulfinate for synthesis. CAS 824-79-3, molar mass 178.19 g/mol., for synthesis

  • CAS Number 824-79-3

  • Linear Formula C₇H₇NaO₂S

  • Molecular Weight 178.19 g/mol

  •  EC Number 212-538-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8436770100)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 95.0 % (a/a)
  Water (K. F.): ≤ 5.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
RTECS 
XT4725000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?