• USA Home
  • 8.43722 - Tetramethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
8.43722 EMD Millipore

Tetramethylammonium bromide

Tetramethylammonium bromide for synthesis. CAS 64-20-0, pH 5.5 - 6.5 (100 g/l, H₂O)., for synthesis

  • CAS Number 64-20-0

  • Linear Formula C₄H₁₂BrN

  • Molecular Weight 154.04 g/mol

  •  EC Number 200-581-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8437220025)
density   1.50 g/cm3
mp   230 °C (decomposition)
pH   5.5 - 6.5 (100 g/l, H₂O)
bulk density   450 kg/m3
LD 50 oral   LD50 Rat 50 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Hazard Statement(s) 
H300: Fatal if swallowed.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
BS7600000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
toxic
R Phrase 
R 25
Toxic if swallowed.
S Phrase 
S 24/25-37-45
Avoid contact with skin and eyes.Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?